MilliporeSigma
  • Metformin sensitizes endometrial cancer cells to chemotherapy by repressing glyoxalase I expression.
Metformin sensitizes endometrial cancer cells to chemotherapy by repressing glyoxalase I expression.
The journal of obstetrics and gynaecology research (2012-05-01)
Lingling Dong, Qian Zhou, Zhenbo Zhang, Yaping Zhu, Tao Duan, Youji Feng
ABSTRACT
Metformin plays an important role in the inhibition of cancer cell growth and prolongs remission durations. It reverses progestin-resistance in endometrial cancer cells by downregulating glyoxalase I (GloI) expression. This study aimed to investigate the effect of metformin on endometrial cancer cell chemotherapeutic sensitivity and explore the underlying molecular mechanisms. MTT assay was performed to determine the rate of cell death after cisplatin and paclitaxel with or without metformin. Western blot was carried out to analyze GloI expression. SiRNA-targeting of GloI was used to knockdown GloI expression before further treatment with chemotherapeutic agents to examine the effect of GloI downregulation on chemotherapy-induced cell killing. In addition, plasmid transfection was used to overexpress GloI and determine whether high GloI levels blocked metformin-enhanced cell sensitivity to chemotherapy. PCR was used to analyze the efficiency of RNA interference and plasmid transfection. The addition of metformin enhanced the sensitivity of endometrial cells to cisplatin and paclitaxel, which was associated with reduced levels of GloI expression. Moreover, low-dose chemotherapeutic drugs alone could not significantly reduce GloI expression, whereas the addition of metformin potently downregulated GloI protein levels. Cisplatin and paclitaxel markedly inhibited the proliferative ability of GloI-depleted endometrial cancer cells. However, the overexpression of GloI abolished the effect of metformin-enhanced cell sensitivity to chemotherapeutic drugs. Metformin enhances the rate of cell-killing induced by chemotherapeutic agents by repressing GloI expression.
Sigma-Aldrich
Glyoxalase I from Saccharomyces cerevisiae, Grade IV, buffered aqueous glycerol solution, ≥400 units/mg protein
Pricing and availability is not currently available.