MilliporeSigma
  • Simultaneous visualization of both signaling cascade activity and end-point gene expression in single cells.
Simultaneous visualization of both signaling cascade activity and end-point gene expression in single cells.
PloS one (2011-06-08)
Irene Weibrecht, Ida Grundberg, Mats Nilsson, Ola Söderberg
ABSTRACT
We have developed an approach for simultaneous detection of individual endogenous protein modifications and mRNA molecules in single cells in situ. For this purpose we combined two methods previously developed in our lab: in situ proximity ligation assay for the detection of individual protein interactions and -modifications and in situ detection of single mRNA molecules using padlock probes. As proof-of-principle, we demonstrated the utility of the method for simultaneous detection of phosphorylated PDGFRβ and DUSP6/MKP-3 mRNA molecules in individual human fibroblasts upon PDGF-BB stimulation. Further we applied drugs disrupting the PDGFRβ signaling pathway at various sites to show that this combined method can concurrently monitor the molecular effect of the drugs, i.e. inhibition of downstream signaling from the targeted node in the signaling pathway. Due to its ability to detect different types of molecules in single cells in situ the method presented here can contribute to a deeper understanding of cell-to-cell variations and can be applied to e.g. pinpoint effector sites of drugs in a signaling pathway.
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Paraformaldehyde, CP
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
L-Cysteine, 97%
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester, ≥98%, powder
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%