MilliporeSigma
  • Effect of triiodothyronine on reactive oxygen species generation by leukocytes, indices of oxidative damage, and antioxidant reserve.
Effect of triiodothyronine on reactive oxygen species generation by leukocytes, indices of oxidative damage, and antioxidant reserve.
Metabolism: clinical and experimental (2000-07-06)
C H Magsino, W Hamouda, H Ghanim, R Browne, A Aljada, P Dandona
ABSTRACT
We have examined the effect of short-term triiodothyronine (T3) administration on reactive oxygen species (ROS) generation by leukocytes in 9 euthyroid subjects. At a dose of 60 microg/d orally for 7 days, T3 induced a significant increase in ROS generation by mononuclear cells (MNCs) from 183 +/- 102 mV at baseline to 313 +/- 111 mV on the seventh day (P < .02), and by polymorphonuclear leukocytes (PMNLs) from 195 +/- 94 mV at baseline to 302 +/- 104 mV on the seventh day (P < .02). There was also a significant increase in meta-tyrosine (P < .001) and ortho-tyrosine (P < .001), known indices of oxidative damage to proteins and amino acids. However, there was no increase in plasma thiobarbituric acid-reactive substances (TBARS), an index of oxidative damage to lipids, and in the level of carbonylated proteins, a less sensitive index to assess protein oxidation. There was no decrease in the level of antioxidants such as alpha-tocopherol, vitamin A, beta-carotene, lycopene, and lutein/zeaxanthin. The stimulatory effect on ROS generation may reflect a generalized increase in metabolic activity or may be a specific effect on NADPH oxidase in leukocyte membranes. The absence of a significant change in TBARS, carbonylated proteins, alpha-tocopherol, vitamin A, beta-carotene, lycopene, and lutein/zeaxanthin may reflect the short duration of the increased ROS load.
Sigma-Aldrich
DL-o-Tyrosine, ≥96.0% (NT)
Pricing and availability is not currently available.