MilliporeSigma
Defensive chemistry of Senecio miser.
Journal of natural products (2001-02-15)
M Reina, A González-Coloma, C Gutiérrez, R Cabrera, M L Rodríguez, V Fajardo, L Villarroel
ABSTRACT
Three eremophilanolides, 1alpha-acetoxy-8beta-methoxy-10betaH-eremophil-7(11)-en-8alpha,12-olide (1); 1alpha-angeloyloxy-6beta-hydroxy-8beta-methoxy-10betaH-eremophil-7(11)-en-8alpha,12-olide (2); and 1alpha-angeloyloxy-8betaH,10betaH-eremophil-7(11)-en-8alpha,12-olide (3), and two pyrrolizidine alkaloids, integerrimine (4) and its N-oxide (5), were isolated from bioactive fractions of Senecio miser. The structures of the new compounds 1 and 2 were established by NMR spectroscopic analysis and chemical transformation. The X-ray analysis of compound 1 was also performed. Eremophilanolides 1 and 2 and alkaloids 4 and 5 were found to be strong insect antifeedants, further supporting a proposed defensive role for these classes of compounds.