MilliporeSigma
  • 3-methylthiopropionaldehyde as precursor of dimethyl trisulfide in aged beers.
3-methylthiopropionaldehyde as precursor of dimethyl trisulfide in aged beers.
Journal of agricultural and food chemistry (2001-04-21)
L Gijs, P Perpète, A Timmermans, S Collin
ABSTRACT
Hop S-methylcysteine sulfoxide has previously been postulated as the precursor of dimethyl trisulfide (DMTS) in beers. The present data point to 3-methylthiopropionaldehyde, the Strecker aldehyde issued from methionine, as another potential precursor in aged beers. Spiking either fresh beer or wort before boiling leads in all cases to higher levels of DMTS after storage. Moreover, special malts with a high level of 3-methylthiopropionaldehyde also favor polysulfide synthesis. A higher pH should increase this onion-like off-flavor, whereas a low pH is unfortunately known to enhance the cardboard flavor of aged beers. 3-methylthiopropanol, issued from yeast reducing activity, can be considered as an additional DMTS source during aging.
Sigma-Aldrich
Dimethyl trisulfide, ≥98%, FG
Pricing and availability is not currently available.
Supelco
Dimethyl trisulfide, analytical standard
Pricing and availability is not currently available.