MilliporeSigma
  • Time-weighted average sampling of airborne n-valeraldehyde by a solid-phase microextration device.
Time-weighted average sampling of airborne n-valeraldehyde by a solid-phase microextration device.
Journal of chromatography. A (2002-06-13)
S W Tsai, T A Chang
ABSTRACT
A solid-phase microextraction (SPME) device was used as a time-weighted average sampler for n-valeraldehyde. The SPME device was first modified to improve the wearer's acceptance as a passive sampler. Then a poly(dimethylsiloxane)-divinylbenzene fiber was used and O-2,3,4,5,6-(pentafluorobenzyl)hydroxylamine hydrochloride (PFBHA) was loaded onto the fiber. Vapors of known concentrations around the threshold limit values time-weighted average of n-valeraldehyde and specific relative humidities (RHs) were generated by syringe pumps in a dynamic generation system. n-Valeraldehyde vapors in gas bags were also generated. An exposure chamber was designed to allow measurement of face velocities, temperatures, exposing vapor concentrations, and RHs. Gas chromatography with flame ionization detection was used for sample analysis. The appropriate adsorption time for SPME coating PFBHA was determined to be 2 min and the desorption time for oxime formed after sampling was optimized to be 2 min. The experimental sampling constant was found to be (3.86+/-0.13) x 10(-2) cm3/min and face velocity was not expect to have effect on the sampler.
Sigma-Aldrich
Valeraldehyde, ≥97%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Valeraldehyde, 97%
Pricing and availability is not currently available.