MilliporeSigma
  • Nitrosation of 2-acetylcyclohexanone. 2. Reaction in water in the absence and presence of cyclodextrins.
Nitrosation of 2-acetylcyclohexanone. 2. Reaction in water in the absence and presence of cyclodextrins.
The Journal of organic chemistry (2003-03-29)
Emilia Iglesias
ABSTRACT
The kinetic study of the nitrosation of the enol of 2-acetylcyclohexanone (ACHE) has been performed in aqueous acid media in the absence and presence of alpha- and beta-cyclodextrin. The reaction is first-order with respect to both reactants concentration: [nitrite] and [ACHE]; but, unexpectly, the dependence of both [H(+)] or [X(-)] (X(-) = Cl(-), Br(-), or SCN(-)) is not simple first-order. The experimental findings have been explained on the basis of a reaction mechanism that considers the formation of a chelate-nitrosyl complex intermediate in steady-state. Addition of both alpha-cyclodextrin (alpha-CD) or beta-cyclodextrin (beta-CD) diminishs strongly the observed rate constant, k(o), measured either for enol-nitrosation or for enol-ketonization reactions. In the case of beta-CD, the inhibition effect is explained through the formation of nonproductive inclusion complexes between the enol (EH) and beta-CD of 1:1 stoichiometry. Nevertheless, the quantitative interpretation of k(o)-[alpha-CD] profiles requires the assumption that the inclusion complexes formation of both 1:1 (EH/alpha-CD) and 1:2 (EH/alpha-CD(2)) stoichiometries. In the case of enol-ketonization, the EH/alpha-CD complex is nearly as reactive as the uncomplexed enol.
Sigma-Aldrich
2-Acetylcyclohexanone, 97%
Pricing and availability is not currently available.