MilliporeSigma
  • Nereistoxin and cartap neurotoxicity attributable to direct block of the insect nicotinic receptor/channel.
Nereistoxin and cartap neurotoxicity attributable to direct block of the insect nicotinic receptor/channel.
Journal of agricultural and food chemistry (2003-04-17)
Seog-Jong Lee, Motohiro Tomizawa, John E Casida
ABSTRACT
Nereistoxin (NTX) (4-dimethylamino-1,2-dithiolane) is the naturally occurring prototype for cartap [the bis(thiocarbamate) derivative of the NTX dithiol], which is generally regarded as a proinsecticide reverting to NTX. The aim of this study is to define the target site(s) for dithiolanes and dithiol esters. The affinity of [(3)H]NTX was not suitable for binding assays with honeybee (Apis mellifera) head membranes. However, NTX and cartap are equally potent, direct-acting, and competitive displacers of [(3)H]thienylcyclohexylpiperidine binding at the noncompetitive blocker (NCB) site of the Apis nicotinic acetylcholine receptor (nAChR)/channel. NTX also binds at the Apis [(3)H]imidacloprid agonist site, but cartap does not. As candidate metabolic pathways, sequential N-desmethylation and S-oxidation of NTX progressively reduce its potency at the NCB site and toxicity to houseflies. A P450 inhibitor reduces the toxicity of NTX and enhances it with cartap. Surprisingly, cartap is not just a pro-NTX but instead directly induces inhibitory neurotoxicity by blocking the nAChR/channel, whereas NTX may have dual NCB and agonist targets.
Product Description
Supelco
Cartap hydrochloride, PESTANAL®, analytical standard
Pricing and availability is not currently available.