MilliporeSigma
  • [Synthesis of a novel starch derivative and its adsorption property for creatinine].
[Synthesis of a novel starch derivative and its adsorption property for creatinine].
Yao xue xue bao = Acta pharmaceutica Sinica (2003-07-02)
Jiu-gao Yu, Dong-zhi Yang
ABSTRACT
To prepare a novel creatinine adsorbent with anti-renal failure activity. A novel starch ester was prepared under heterogeneous condition by the reaction of starch (ST) and 3, 5-dinitrobenzoyl chloride (DNBZ-Cl). The products were characterized by means of elemental analysis, FTIR, 13CNMR and UV-Vis spectra. The adsorption properties of DNBZ-ST were examined. The adsorption equilibrium is reached after 4 h, the adsorption capacity increases with the degree of substitution (DS) of adsorbent and creatinine concentration. When creatinine concentration is higher than 300 mg.L-1, concentration showed no apparent effect on adsorption capacity. As the adsorption temperature varied from 19 degrees C to 49 degrees C, adsorption capacity first decreases and then increases, and is lowest at 37 degrees C. The adsorption capacity first increases and then decreases as the pH value increase from 4 to 11 and is highest at pH 8. The highest adsorption capacity reached 25 mg.g-1 at 37 degrees C pH 7 and the creatinine concentration is 100 mg.L-1. Starch 3,5-dinitrobenzoate showed a better adsorption property for creatinine, and it is worthy to be studied more deeply.
Sigma-Aldrich
3,5-Dinitrobenzoyl chloride, ≥96.5%
Pricing and availability is not currently available.