MilliporeSigma
  • Simultaneous determination of fatty, dicarboxylic and amino acids based on derivatization with isobutyl chloroformate followed by gas chromatography-positive ion chemical ionization mass spectrometry.
Simultaneous determination of fatty, dicarboxylic and amino acids based on derivatization with isobutyl chloroformate followed by gas chromatography-positive ion chemical ionization mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2003-12-31)
Tim G Sobolevsky, Alexander I Revelsky, Igor A Revelsky, Barbara Miller, Vincent Oriedo
ABSTRACT
Gas chromatography-mass spectrometry (GC-MS) with positive ion chemical ionization (PICI) using isobutane as reagent gas was applied for analysis of isobutoxycarbonyl/isobutyl derivatives of 13 fatty, 6 dicarboxylic and 13 amino acids in a single run. For all investigated compounds (except several amino acids) the quasimolecular ions [MH](+) were registered. Asparagine underwent fragmentation via decarboxylation followed by elimination of OC(4)H(9) ([M-117](+)), whereas serine and tyrosine produced the cluster ions [M+C(4)H(9)OCO](+). Estimated detection limits were 6-250 pg in the total ion current (TIC) mode and 3-10 times lower using the selected-ion monitoring (SIM) mode.
Sigma-Aldrich
Isobutyl chloroformate, 98%
Pricing and availability is not currently available.