MilliporeSigma
  • Total degradation of pentachloroethane by an engineered Alcaligenes strain expressing a modified camphor monooxygenase and a hybrid dioxygenase.
Total degradation of pentachloroethane by an engineered Alcaligenes strain expressing a modified camphor monooxygenase and a hybrid dioxygenase.
Bioscience, biotechnology, and biochemistry (2004-06-25)
Ryo Iwakiri, Kunichika Yoshihira, Ngadiman, Taiki Futagami, Masatoshi Goto, Kensuke Furukawa
ABSTRACT
We engineered biphenyl-degrading Alcaligenes sp. strain KF711 for total degradation of pentachloroethane (PCA), which expresses a modified camphor monooxygenase and a hybrid dioxygenase consisting of TodC1 (a large subunit of toluene dioxygenase of Pseudomonas putida F1) and BphA2-BphA3-pbhA4 (a small subunit, ferredoxin and ferredoxin reductase of biphenyl dioxygenase, respectively, in strain KF707). Modified camphor monooxygenase genes (camCAB) were supplied as a plasmid and the todC1 gene was integrated within the chromosomal bph gene cluster by a single crossover recombination. The resultant strain KF711S-3cam dechlorinated PCA to trichloroethene by the action of the modified camphor monooxygenase under anaerobic conditions. The same strain subsequently degraded trichloroethene formed oxidatively by the action of the Tol-Bph hybrid dioxygenase under aerobic conditions. Thus sequential anaerobic and aerobic treatments of the KF711S-3cam resting cells resulted in efficient and total degradation of PCA.
Sigma-Aldrich
Pentachloroethane, 95%
Pricing and availability is not currently available.