MilliporeSigma
  • Effect of mobile phase composition, pH and buffer type on the retention of ionizable compounds in reversed-phase liquid chromatography: application to method development.
Effect of mobile phase composition, pH and buffer type on the retention of ionizable compounds in reversed-phase liquid chromatography: application to method development.
Journal of chromatography. A (2004-10-16)
S Heinisch, J L Rocca
ABSTRACT
Optimizing separation of ionizable compounds in order to find robust conditions has become an important part of method development in liquid chromatography. This work is an attempt to explain the observed variations of retention of acid and basic compounds with the organic modifier content in the mobile phase, according to various factors: the type of modifier, the type of buffer, the temperature and of course the type of solute. This is done by considering the variation of the so-called chromatographic pKa which refers to the pH measured in the aqueous medium and is determined from retention data. A procedure is described that accurately relates, from nine experiments, retention to solvent composition and pH. The limits of such a procedure are evaluated and two examples of optimized separations of basic compounds are given.
Sigma-Aldrich
4-Nitrobenzoic acid, purum, ≥98.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Nitrobenzoic acid, 98%
Pricing and availability is not currently available.