MilliporeSigma
  • Dynamic nuclear polarization properties of nitroxyl radicals used in Overhauser-enhanced MRI for simultaneous molecular imaging.
Dynamic nuclear polarization properties of nitroxyl radicals used in Overhauser-enhanced MRI for simultaneous molecular imaging.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2006-08-01)
A Milton Franklin Benial, Kazuhiro Ichikawa, Ramachandran Murugesan, Ken-ichi Yamada, Hideo Utsumi
ABSTRACT
DNP parameters relevant to Overhauser-enhanced magnetic resonance imaging (OMRI) are reported for a few nitroxyl radicals and their corresponding (15)N and (2)H enriched analogues, used in simultaneous imaging by OMRI. DNP enhancement was measured at 14.529 mT, using a custom-built scanner operating in a field-cycled mode, for different concentrations, ESR irradiation times and RF power levels. DNP enhancements increased with agent concentration up to 2.5 mM and decreased above 3 mM, in tune with ESR line broadening measured at X-band as a function of the agent concentration. The proton spin-lattice relaxation times (T(1)) measured at very low Zeeman field (14.529 mT) and the longitudinal relaxivity parameters were estimated. The relaxivity parameters were in good agreement with those independently computed from the linear region of the concentration dependent enhancement. The leakage factor showed an asymptotic increase with increasing agent concentration. The coupling parameters of (14)N- and (15)N-labeled carbamoyl-PROXYL showed the interaction between the electron and nuclear spins to be mainly dipolar in origin. Upon (2)H labeling, about 70% and 40% increases in enhancement for (15)N- and (14)N-labeled nitroxyl agents were observed, respectively. It is envisaged that the results reported here may enable better understanding of the factors determining DNP enhancement to design suitable 'beacons' for simultaneous molecular imaging by OMRI.
Sigma-Aldrich
3-Carbamoyl-PROXYL
Pricing and availability is not currently available.