MilliporeSigma
  • Effect of lactoferrin on the growth performance, intestinal morphology, and expression of PR-39 and protegrin-1 genes in weaned piglets.
Effect of lactoferrin on the growth performance, intestinal morphology, and expression of PR-39 and protegrin-1 genes in weaned piglets.
Journal of animal science (2006-09-15)
Y Wang, T Shan, Z Xu, J Liu, J Feng
ABSTRACT
A total of 90 weaned female pigs (Duroc x Landrace x Yorkshire) were used in a 15-d growth experiment to investigate the effect of lactoferrin on growth performance, intestinal morphology, and expression of PR-39 and protegrin-1 genes. The pigs were allocated on the basis of BW and litter to 3 dietary treatments in a randomized complete block design. There were 3 replicate pens per treatment, and the pigs were grouped with 10 pigs per pen. The dietary treatments were (1) basal diet; (2) basal diet + 20 mg of flavomycin/kg + 110 mg of aureomycin/kg; (3) basal diet + 1.0 g of lactoferrin/kg. Six pigs, randomly selected from each treatment (2 piglets/pen) were slaughtered for intestinal morphology and expression of PR-39 and protegrin-1 genes at the end of the experiment. Supplementation with lactoferrin improved growth performance; it increased ADG by 41.80% (P < 0.01) and efficiency of gain (G:F) by 17.20% (P < 0.05). Intestinal villus height was increased by 15.30% (P < 0.05), and crypt depth was decreased by 9.60% (P < 0.05). Supplemental lactoferrin increased the relative abundance of mRNA for PR-39 and protegrin-1 by 143% (P < 0.01) and 217% (P < 0.01), respectively. The use of lactoferrin as an additive to improve nonspecific immunity and strengthen host defenses would be good a method of defending weaned pigs from infections and weanling stress.