MilliporeSigma
  • Investigation of the excited state structure of DCM via ultrafast electronic pump/vibrational probe.
Investigation of the excited state structure of DCM via ultrafast electronic pump/vibrational probe.
The journal of physical chemistry. B (2006-09-22)
Aaron J Van Tassle, Matthew A Prantil, Graham R Fleming
ABSTRACT
Time resolved visible pump, infrared probe transient absorption measurements of the solutes 4-dicyanomethylene-2-methyl-6-(p-(dimethylamino)styryl)-4H-pyran (DCM) and its isotopomer DCM-d6 are employed to probe the dynamics of charge transfer state formation in dimethyl sulfoxide (DMSO) and acetonitrile (MeCN). We observe a two stage charge transfer (CT): the first step is an instrument-response-limited charge separation to the dicyanomethylene group, and the second involves a structural evolution of the dimethylamino group. Theoretical calculations and isotopic substitution indicate that the observed vibration is due to the dimethylamino group twisting out of plane, stabilizing the charge separation.
Sigma-Aldrich
4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran, Dye content 98 %
Pricing and availability is not currently available.