MilliporeSigma
  • Rapid determination of protopine, allocryptopine, sanguinarine and chelerythrine in fruits of Macleaya cordata by microwave-assisted solvent extraction and HPLC-ESI/MS.
Rapid determination of protopine, allocryptopine, sanguinarine and chelerythrine in fruits of Macleaya cordata by microwave-assisted solvent extraction and HPLC-ESI/MS.
Phytochemical analysis : PCA (2006-12-06)
Xu-Biao Luo, Bo Chen, Shou-Zhuo Yao
ABSTRACT
An isocratic high-performance liquid chromatographic method coupled with electrospray mass spectrometry was developed to determine protopine, allocryptopine, sanguinarine and chelerythrine in fruits of Macleaya cordata. The sample was extracted with hydrochloric acid aqueous solution using microwave-assisted extraction method. The extracts were separated on a C8 reversed-phase HPLC column with acetonitrile:acetate buffer as mobile phase, and full elution of all analytes was realized isocratically within 10 min. The abundance of pseudomolecule ions was recorded using selected ion recording at m/z 354.4, 370.1, 332.5, 348.5 and 338.5 for protopine, allocryptopine, sanguinarine, chelerythrine and the internal standard, jatrorrhizine, respectively. Internal standard curves were used for the quantification of protopine, allocryptopine, sanguinarine and chelerythrine, which showed a linear range of 0.745-74.5, 0.610-61.0, 0.525-105 and 0.375-75 microg/mL, respectively, with correlation coefficients of 0.9995, 0.9992, 0.9993 and 0.9989, and limits of detection of 3.73, 3.05, 1.60 and 1.11 ng/mL, respectively.
Sigma-Aldrich
Protopine hydrochloride, ≥98%, solid
Pricing and availability is not currently available.