MilliporeSigma
  • Fucosylated pentaerythrityl phosphodiester oligomers (PePOs): automated synthesis of DNA-based glycoclusters and binding to Pseudomonas aeruginosa lectin (PA-IIL).
Fucosylated pentaerythrityl phosphodiester oligomers (PePOs): automated synthesis of DNA-based glycoclusters and binding to Pseudomonas aeruginosa lectin (PA-IIL).
Bioconjugate chemistry (2007-07-31)
François Morvan, Albert Meyer, Anne Jochum, Charles Sabin, Yann Chevolot, Anne Imberty, Jean-Pierre Praly, Jean-Jacques Vasseur, Eliane Souteyrand, Sébastien Vidal
ABSTRACT
The synthesis of propargylated pentaerythrityl phosphodiester oligomers (PePOs) was achieved using a DNA synthesizer with a bis-propargylated pentaerythritol-based phosphoramidite. An azido fucose derivative was reacted under "click" chemistry conditions activated by microwaves to construct a series of glycosylated PePOs bearing 4, 6, 8, and 10 L-fucose residues. Binding to the fucose-specific bacterial lectin (PA-IIL) was determined for the fucosylated PePOs through an enzyme-linked lectin amplification competition assay. The IC50 values measured are 10-20 times better than for monovalent l-fucose and denotate for a "macromolecular" effect rather than a "cluster" effect.
Sigma-Aldrich
Pentaerythritol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Pentaerythritol, 98%
Pricing and availability is not currently available.