MilliporeSigma
  • Tissues distribution of R-(-)- and S-(+)-m-nisoldipine after single enantiomer administration in rats.
Tissues distribution of R-(-)- and S-(+)-m-nisoldipine after single enantiomer administration in rats.
Drug development and industrial pharmacy (2008-09-30)
Min Li, Qiao Wang, Chunying Wang, Xiujuan Jing, Kunfeng Duan, Xiang'ai Chen, Lei Xu, Yaping Tian, Lantong Zhang, Yumin Du, Xiaowei Zhang, Xiaona Sheng
ABSTRACT
Rapid, sensitive, and selective high-performance liquid chromatography methods were developed and validated for determination of m-nisoldipine enantiomers in rat tissues. All of the samples were prepared based on a simple and efficient liquid-liquid extraction method. After validating that no racemation occurred by ULTRON ES-OVM (Japan), m-nisoldipine enantiomers were determined, respectively, on a reverse-phase C(18) column (5 microm, 250 x 4.6 mm). This method was applied to study tissue distribution of m-nisoldipine enantiomers in rats after a single administration of m-nisoldipine enantiomers. By the two-sample t test, there were basically no significant differences between the two enantiomers in each tissue ( p > .05), which indicates that they may have the same potency in rats. In small intestine, lung, liver, and spleen, the concentrations of R-(-)- and S-(+)-m-nisoldipine were high at 30 and 150 min than that at 90 min, which showed that m-nisoldipine enantiomers may have the phenomenon of hepatoenteral circulation. A small quantity of the prototype of R-(-)- and S-(+)-m-nisoldipine in brain showed that they can cross the blood-brain barrier to arrive at the brain tissue. The high quantity of distribution in lung and brain implied that the lipophilicity of the drug was powerful.
Sigma-Aldrich
Nisoldipine, ≥98% (HPLC)
Pricing and availability is not currently available.