MilliporeSigma
  • Fabrication of biomimetic superhydrophobic surface on engineering materials by a simple electroless galvanic deposition method.
Fabrication of biomimetic superhydrophobic surface on engineering materials by a simple electroless galvanic deposition method.
Langmuir : the ACS journal of surfaces and colloids (2009-12-17)
Xianghui Xu, Zhaozhu Zhang, Jin Yang
ABSTRACT
We have reported an easy means in this paper to imitate the "lotus leaf" by constructing a superhydrophobic surface through a process combining both electroless galvanic deposition and self-assembly of n-octadecanethiol. Superhydrophobicity with a static water contact angle of about 169 +/- 2 degrees and a sliding angle of 0 +/- 2 degrees was achieved. Both the surface chemical compositions and morphological structures were analyzed. We have obtained a feather-like surface structure, and the thickness of the Ag film is about 10-30 microm. The stability of the superhydrophobic surface was tested under the following three conditions: (1) pH value from 1 to 13; (2) after freezing treatment at -20 degrees C; (3) at ambient temperature. It shows a notable stability in that the contact angle of the sample still remained higher than 150 degrees in different conditions. It can be concluded that our approach can provide an alternative way to fabricate stable superhydrophobic materials.
Sigma-Aldrich
1-Octadecanethiol, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Octadecanethiol, purum, ≥95.0% (GC)
Pricing and availability is not currently available.