MilliporeSigma
  • Enduracidin analogues with altered halogenation patterns produced by genetically engineered strains of Streptomyces fungicidicus.
Enduracidin analogues with altered halogenation patterns produced by genetically engineered strains of Streptomyces fungicidicus.
Journal of natural products (2010-04-01)
Xihou Yin, Ying Chen, Ling Zhang, Yang Wang, T Mark Zabriskie
ABSTRACT
Enduracidins (1, 2) and ramoplanin (3) are structurally and functionally closely related lipodepsipeptide antibiotics. They are active against multi-drug-resistant Gram-positive pathogens, including MRSA. Each peptide contains one chlorinated non-proteinogenic amino acid residue, Cl(2)-Hpg or Cl-Hpg. To investigate the timing of halogenation and the importance of chlorination on bioactivity and bioavailability of enduracidin, and to probe the substrate specificity and portability of the ramoplanin halogenase, we constructed the mutant strain SfDelta30 in which the enduracidin halogenase gene orf30 had been deleted and complemented it with the ramoplanin counterpart orf20. We also expressed orf20 in the enduracidin wild-type producer. Metabolite analysis revealed SfDelta30 produced the novel analogues dideschloroenduracidins A (4) and B (5), while the recombinant strains SfDelta30R20 and SfR20 produced monodeschloroenduracidins A (6) and B (7) and a trichlorinated enduracidin (8), respectively. In addition, orf30 self-complementation yielded the strain SfDelta30E30, which is capable of producing six peptides including 6 and 7. MS/MS analysis positioned the single chlorine atom in 6 at Hpg(13) and localized the third chlorine atom in 8 to Hpg(11). Biological evaluation of these enduracidin analogues indicated that all retained activity against Staphylococcus aureus. Our findings lay the foundation for further utilization of enduracidin and ramoplanin halogenases in combinatorial biosynthesis.
Sigma-Aldrich
Ramoplanin
Pricing and availability is not currently available.