MilliporeSigma
  • [The synthesis of O-substituted 3-oximes of 6alpha-methyl-16alpha,17alpha-cyclohexanopregn-4-ene-3,20-dione tritium-labeled in 1 and 2 positions].
[The synthesis of O-substituted 3-oximes of 6alpha-methyl-16alpha,17alpha-cyclohexanopregn-4-ene-3,20-dione tritium-labeled in 1 and 2 positions].
Bioorganicheskaia khimiia (2010-06-10)
V P Shevchenko, I Iu Nagaev, I S Levina, L E Kulikova, N F Miasoedov, A V Kamernitzkiĭ
ABSTRACT
1,2-Tritium-labeled 3-(O-carboxypropyl)- and 3-(O-carbomethoxypropyl)-oximes of 6alpha-methyl-16alpha,17alpha-cyclohexanopregn-4-ene-3,20-diones were obtained by the homogeneous catalytic hydrogenation of 1,2-dehydroprecursors with gaseous tritium and the subsequent separation of the resulting mixtures by HPLC. The specific radioactivities of 50-55 Ci/mmol were prepared using tris-(triphenylphosphine)-rhodium chloride.
Sigma-Aldrich
Tris(triphenylphosphine)rhodium(I) chloride
Pricing and availability is not currently available.
Sigma-Aldrich
Tris(triphenylphosphine)rhodium(I) chloride, 99.9% trace metals basis
Pricing and availability is not currently available.