MilliporeSigma
  • Photocrosslinkable chitosan modified with angiopoietin-1 peptide, QHREDGS, promotes survival of neonatal rat heart cells.
Photocrosslinkable chitosan modified with angiopoietin-1 peptide, QHREDGS, promotes survival of neonatal rat heart cells.
Journal of biomedical materials research. Part A (2010-06-12)
Fiona Rask, Susan M Dallabrida, Nesreen S Ismail, Zohreh Amoozgar, Yoon Yeo, Maria A Rupnick, Milica Radisic
ABSTRACT
Myocardial infarction (MI) results in the death of cardiomyocytes (CM), which causes scar formation and pathological remodeling of the heart. The delivery of healthy myocytes or bone marrow cells reduces pathological remodeling after MI, however, current cell injection methods have low cell survival rates and high cell loss. The main objective of this work was to develop a novel hydrogel that can promote survival of CMs. Photocrosslinkable azidobenzoic acid modified chitosan (Az-chitosan) was conjugated with the angiopoietin-1-derived peptide, QHREDGS. This novel peptide is thought to mediate attachment and survival responses of CM to angiopoietin-1 via integrin binding. Thin layers of Az-chitosan, Az-chitosan-QHREDGS, and Az-chitosan-DGQESHR (scrambled peptide control) were spin coated on glass slides and photocrosslinked with application of UV light (365 nm). Neonatal rat heart cells cultured up to 5 days, demonstrated significantly higher attachment and viability on Az-chitosan-QHREDGS compared to cells on other hydrogel controls. Surfaces were also stained for the CM-specific marker troponin I, demonstrating significantly higher percentage of CMs on Az-chitosan-QHREDGS compared to Az-chitosan. The cells cultivated on Az-chitosan-QHREDGS demonstrated significantly lower levels of caspase 3/7 activation after taxol treatment in comparison to cells cultivated on the control hydrogels, glass substrate, or Az-chitosan linked to RGD, an established integrin binding peptide that did not protect against apoptosis. Thus, Az-chitosan-QHREDGS supports attachment and survival of neonatal rat heart cells.
Sigma-Aldrich
4-Azidobenzoic acid solution, ~0.2 M in tert-butyl methyl ether, ≥95.0% (HPLC)
Pricing and availability is not currently available.