MilliporeSigma
  • Images of paraffin monolayer crystals with perfect contrast: minimization of beam-induced specimen motion.
Images of paraffin monolayer crystals with perfect contrast: minimization of beam-induced specimen motion.
Ultramicroscopy (2010-12-28)
R M Glaeser, G McMullan, A R Faruqi, R Henderson
ABSTRACT
Quantitative analysis of electron microscope images of organic and biological two-dimensional crystals has previously shown that the absolute contrast reached only a fraction of that expected theoretically from the electron diffraction amplitudes. The accepted explanation for this is that irradiation of the specimen causes beam-induced charging or movement, which in turn causes blurring of the image due to image or specimen movement. In this paper, we used three different approaches to try to overcome this image-blurring problem in monolayer crystals of paraffin. Our first approach was to use an extreme form of spotscan imaging, in which a single image was assembled on film by the successive illumination of up to 50,000 spots, each of a diameter of around 7 nm. The second approach was to use the Medipix II detector with its zero-noise readout to assemble a time-sliced series of images of the same area in which each frame from a movie with up to 400 frames had an exposure of only 500 electrons. In the third approach, we simply used a much thicker carbon support film to increase the physical strength and conductivity of the support. Surprisingly, the first two methods involving dose fractionation in space or time produced only partial improvements in contrast whereas the third approach produced many virtually perfect images, where the absolute contrast predicted from the electron diffraction amplitudes was observed in the images. We conclude that it is possible to obtain consistently almost perfect images of beam-sensitive specimens if they are attached to an appropriately strong and conductive support; however great care is needed in practice and the problem remains of how to best image ice-embedded biological structures in the absence of a strong, conductive support film.
Sigma-Aldrich
Tetratetracontane, 99%
Pricing and availability is not currently available.
Supelco
Tetratetracontane, analytical standard
Pricing and availability is not currently available.