MilliporeSigma
  • Skin-depigmenting agent monobenzone induces potent T-cell autoimmunity toward pigmented cells by tyrosinase haptenation and melanosome autophagy.
Skin-depigmenting agent monobenzone induces potent T-cell autoimmunity toward pigmented cells by tyrosinase haptenation and melanosome autophagy.
The Journal of investigative dermatology (2011-02-18)
Jasper G van den Boorn, Daisy I Picavet, Paul F van Swieten, Henk A van Veen, Debby Konijnenberg, Peter A van Veelen, Toni van Capel, Esther C de Jong, Eric A Reits, Jan W Drijfhout, Jan D Bos, Cornelis J M Melief, Rosalie M Luiten
ABSTRACT
In this study, we report the previously unknown mechanism of inducing robust anti-melanoma immunity by the vitiligo-inducing compound monobenzone. We show monobenzone to increase melanocyte and melanoma cell immunogenicity by forming quinone-haptens to the tyrosinase protein and by inducing the release of tyrosinase- and melanoma antigen recognized by T cells-1 (MART-1)-containing CD63+ exosomes following melanosome oxidative stress induction. Monobenzone further augments the processing and shedding of melanocyte-differentiation antigens by inducing melanosome autophagy and enhanced tyrosinase ubiquitination, ultimately activating dendritic cells, which induced cytotoxic human melanoma-reactive T cells. These T cells effectively eradicate melanoma in vivo, as we have reported previously. Monobenzone thereby represents a promising and readily applicable compound for immunotherapy in melanoma patients.
Sigma-Aldrich
4-(Benzyloxy)phenol, 98%
Pricing and availability is not currently available.