MilliporeSigma
  • Spectrophotometric determination of trace levels of allura red in water samples after separation and preconcentration.
Spectrophotometric determination of trace levels of allura red in water samples after separation and preconcentration.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-02-26)
Mustafa Soylak, Yunus Emre Unsal, Mustafa Tuzen
ABSTRACT
A new, simple, rapid and sensitive solid phase extraction procedure based on separation and preconcentration of trace levels of allura red in water samples on MCI GEL CHP20P column system has been established. Optimal analytical conditions including pH, amount of allura red, flow rates of sample and eluent solutions, resin amount, sample volumes, etc. were investigated. The interference effect of some cations and anions was also studied. Preconcentration factor was found as 250. The detection limit (3 sigma) of the reagent blank for allura red was 2.35 μg L(-1). The determination of allura red levels in samples was performed by using spectrophotometry at 506 nm. The relative standard deviations of the procedure were below 7%. The validation of the presented procedure was performed by the method of standard addition. The procedure was successfully applied to the determination of trace levels of allura red in tap water and waste water samples.
Product Description
Supelco
Allura Red AC, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Allura Red AC, Dye content 80 %
Pricing and availability is not currently available.