MilliporeSigma
[Studies on flavonoids from the leaves of Lindera aggregata].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2011-08-09)
Mei Xiao, Ning Cao, Jing-jing Fan, Yan Shen, Qiang Xu
ABSTRACT
To investigate the chemical constituents in the leaves of Lindera aggregate. Compounds were separated by column chromatography with silica gel and ODS. The structures were identified by physicochemical properties and spectral analysis (1H-NMR, 13C-NMR). Eight compounds were isolated and identified as: quercetin-3-O-L-rhamnoside(1), kaempferol-3-O-L-rhamnoside(2), kaempferol(3), dihydrokaempferol-3-O-L-rhamnoside (4), quercetin (5), quercetin-3-O-alpha-D-arabinofuranoside (6), quercetin-3-O-alpha-D-glucopyranoside(7), kaempferol-3-O-D-glucopyranoside(8). Compounds 2,4 and 8 are obtained from Lindera aggregata for the first time.
Sigma-Aldrich
Kaempferol 3-β-D-glucopyranoside, ≥97.0% (HPLC)
Pricing and availability is not currently available.