MilliporeSigma
  • Atomic and electronic structure of the BaTiO3/Fe interface in multiferroic tunnel junctions.
Atomic and electronic structure of the BaTiO3/Fe interface in multiferroic tunnel junctions.
Nano letters (2011-12-24)
Laura Bocher, Alexandre Gloter, Arnaud Crassous, Vincent Garcia, Katia March, Alberto Zobelli, Sergio Valencia, Shaïma Enouz-Vedrenne, Xavier Moya, Neil D Mathur, Neil D Marthur, Cyrile Deranlot, Stéphane Fusil, Karim Bouzehouane, Manuel Bibes, Agnès Barthélémy, Christian Colliex, Odile Stéphan
ABSTRACT
Artificial multiferroic tunnel junctions combining a ferroelectric tunnel barrier of BaTiO(3) with magnetic electrodes display a tunnel magnetoresistance whose intensity can be controlled by the ferroelectric polarization of the barrier. This effect, called tunnel electromagnetoresistance (TEMR), and the corollary magnetoelectric coupling mechanisms at the BaTiO(3)/Fe interface were recently reported through macroscopic techniques. Here, we use advanced spectromicroscopy techniques by means of aberration-corrected scanning transmission electron microscopy (STEM) and electron energy-loss spectroscopy (EELS) to probe locally the nanoscale structural and electronic modifications at the ferroelectric/ferromagnetic interface. Atomically resolved real-space spectroscopic techniques reveal the presence of a single FeO layer between BaTiO(3) and Fe. Based on this accurate description of the studied interface, we propose an atomistic model of the ferroelectric/ferromagnetic interface further validated by comparing experimental and simulated STEM images with atomic resolution. Density functional theory calculations allow us to interpret the electronic and magnetic properties of these interfaces and to understand better their key role in the physics of multiferroics nanostructures.
Sigma-Aldrich
Barium titanate(IV), powder, <3 μm, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Barium titanate(IV), powder, <2 μm, 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Barium titanate(IV), nanopowder (cubic crystalline phase), <100 nm particle size (BET), ≥99% trace metals basis
Pricing and availability is not currently available.