MilliporeSigma
  • 18-hydroxycorticosterone, 18-hydroxycortisol, and 18-oxocortisol in the diagnosis of primary aldosteronism and its subtypes.
18-hydroxycorticosterone, 18-hydroxycortisol, and 18-oxocortisol in the diagnosis of primary aldosteronism and its subtypes.
The Journal of clinical endocrinology and metabolism (2012-01-13)
Paolo Mulatero, Stefania Morra di Cella, Silvia Monticone, Domenica Schiavone, Maria Manzo, Giulio Mengozzi, Franco Rabbia, Massimo Terzolo, Elise P Gomez-Sanchez, Celso E Gomez-Sanchez, Franco Veglio
ABSTRACT
Diagnosis of primary aldosteronism (PA) is made by screening, confirmation testing, and subtype diagnosis (computed tomography scan and adrenal vein sampling). However, some tests are costly and unavailable in most hospitals. The aim of the study was to evaluate the role of serum 18-hydroxycorticosterone (s18OHB), urinary and serum 18-hydroxycortisol (u- and s18OHF), and urinary and serum 18-oxocortisol (u- and s18oxoF) in the diagnosis of PA and its subtypes, aldosterone-producing adenoma (APA) and bilateral adrenal hyperplasia (BAH). The study included 62 patients with low-renin essential hypertension (EH), 81 patients with PA (20 APA, 61 BAH), 24 patients with glucocorticoid-remediable aldosteronism, 16 patients with adrenal incidentaloma, and 30 normotensives. We measured s18OHB, s18OHF, and s18oxoF before and after saline load test (SLT) and 24-h u18OHF and u18oxoF. PA patients displayed significantly higher levels of s18OHB, u18OHF, and u18oxoF compared to EH and normal subjects; APA patients displayed s18OHB, u18OHF, and u18oxoF levels significantly higher than BAH patients. Similar results were obtained for s18OHF and s18oxoF. SLT significantly reduced s18OHB, s18OHF, and s18oxoF in all groups, but steroid reduction was much less for APA patients compared to BAH and EH. The s18OHB/aldosterone ratio after SLT more than doubled in EH but remained unchanged in APA patients. u18OHF, u18oxoF, and s18OHB measurements in patients with a positive aldosterone/plasma renin activity ratio correlate with confirmatory tests and adrenal vein sampling in PA patients. If verified, these steroid assays would refine the diagnostic workup for PA.
Sigma-Aldrich
18-Hydroxycorticosterone, 97% (CP)
Pricing and availability is not currently available.