MilliporeSigma
Leucobacter denitrificans sp. nov., isolated from cow dung.
Journal of microbiology (Seoul, Korea) (2012-03-01)
Hang-Yeon Weon, Rangasamy Anandham, Tomohiko Tamura, Moriyuki Hamada, Soo-Jin Kim, Yi-Seul Kim, Ken-Ichiro Suzuki, Soon-Wo Kwon
ABSTRACT
The bacterial strain M1T8B10(T) was isolated from cow dung in Suwon, Republic of Korea. The strain was a Gram stain-positive rod, nonmotile, and non-spore-forming. According to 16S rRNA gene sequence analysis, the strain fell within the clade of the genus Leucobacter, showing the highest sequence similarities with Leucobacter aridicollis L-9(T) (98.7%), Leucobacter iarius 40(T) (98.4%), and Leucobacter komagatae JCM 9414(T) (98.2%). Cell-wall peptidoglycan contained the diagnostic diamino acid 2,4-diaminobutyric acid of the genus Leucobacter, showing B-type cross-linked peptidoglycans. The major fatty acids were anteiso-C(15:0), iso-C(16:0), and anteiso-C(17:0). The quinone system consisted of the menaquinones MK-11 (78%) and MK-10 (22%). The polar lipid profiles contained diphosphatidylglycerol, phosphatidylglycerol, and an unidentified glycolipid. Differences in several physiological features including nitrate reduction enabled the isolate to be differentiated from all recognized Leucobacter species. Based on these phylogenetic, chemotaxonomic, and phenotypic results, the isolate represents a novel species, for which the name Leucobacter denitrificans sp. nov. is proposed. The type strain is M1T8B10(T) (=KACC 14055(T) =NBRC 106309(T)).
Sigma-Aldrich
L-2,4-Diaminobutyric acid dihydrochloride, ≥95.0%
Pricing and availability is not currently available.