MilliporeSigma
  • Involvement of inositol 1,4,5-trisphosphate formation in the voltage-dependent regulation of the Ca(2+) concentration in porcine coronary arterial smooth muscle cells.
Involvement of inositol 1,4,5-trisphosphate formation in the voltage-dependent regulation of the Ca(2+) concentration in porcine coronary arterial smooth muscle cells.
The Journal of pharmacology and experimental therapeutics (2012-05-17)
Hisao Yamamura, Susumu Ohya, Katsuhiko Muraki, Yuji Imaizumi
ABSTRACT
The involvement of inositol 1,4,5-trisphosphate (IP(3)) formation in the voltage-dependent regulation of intracellular Ca(2+) concentration ([Ca(2+)](i)) was examined in smooth muscle cells of the porcine coronary artery. Slow ramp depolarization from -90 to 0 mV induced progressive [Ca(2+)](i) increase. The slope was reduced or increased in the presence of Cd(2+) or (±)-1,4-dihydro-2,6-dimethyl-5-nitro-4-(2-[trifluoromethyl]-phenyl)pyridine-3-carboxlic acid methyl ester (Bay K 8644), respectively. The decrease in [Ca(2+)](i) via the membrane hyperpolarization induced by K(+) channel openers (levcromakalim and Evans blue) under current clamp was identical to that under voltage clamp. The step hyperpolarization from -40 to -80 mV reduced [Ca(2+)](i) uniformly over the whole-cell area with a time constant of ∼10 s. The [Ca(2+)](i) at either potential was unaffected by heparin, an inhibitor of IP(3) receptors. Alternatively, [Ca(2+)](i) rapidly increased in the peripheral regions by depolarization from -80 to 0 mV and stayed at that level (∼400 nM) during a 60-s pulse. When the pipette solution contained IP(3) pathway blockers [heparin, 2-aminoethoxydiphenylborate, xestospongin C, or 1-[6-[((17β)-3-methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]-1H-pyrrole-2,5-dione (U73122)], the peak [Ca(2+)](i) was unchanged, but the sustained [Ca(2+)](i) was gradually reduced by ∼250 nM within 30 s. In the presence of Cd(2+), a long depolarization period slightly increased the [Ca(2+)](i), which was lower than that in the presence of heparin alone. In coronary arterial myocytes, the sustained increase in the [Ca(2+)](i) during depolarization was partly caused by the Ca(2+) release mediated by the enhanced formation of IP(3). The initial [Ca(2+)](i) elevation triggered by the Ca(2+) influx though voltage-dependent Ca(2+) channels may be predominantly responsible for the activation of phospholipase C for IP(3) formation.
Sigma-Aldrich
(S)-(−)-Bay K8644, ≥98% (HPLC), solid
Pricing and availability is not currently available.