MilliporeSigma
  • [A role of D-dimer and fibrinopeptide A in diagnosis of a hemostasis system disorders].
[A role of D-dimer and fibrinopeptide A in diagnosis of a hemostasis system disorders].
Klinichna khirurhiia (2012-05-31)
T I Omarov, G A Sultanov, V S Ragimov
ABSTRACT
The investigations, concerning detection of the hemostasis system activation, were done in 26 patients, suffering various critical morbid states (an acute pancreatitis). The contents of products of the enzymes lysis of coagulation system and fibrinolytic system constitute one of the most precise indices. Fibrinopeptid A (FpA) is considered one of the most secure indices, confirming intravascular thrombin formation, and D-dimer--of a fibrin formation. In the patients examined a trustworthy increase of a D-dimer and FpA contents was registered, witnessing the hemostasis system activation in an acute pancreatitis as well as an excessive formation and lysis of fibrin. D-dimer and FpA contents in a plasma constitutes an important diagnostic index, its determination secures the possibility of early diagnosis and control of a hemostasis system disorders.
Sigma-Aldrich
Fibrinopeptide A human, ≥96% (HPLC)
Pricing and availability is not currently available.