MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • HPLC-fluorescence method for measurement of the uremic toxin indoxyl sulfate in plasma.
HPLC-fluorescence method for measurement of the uremic toxin indoxyl sulfate in plasma.
Journal of chromatographic science (2012-06-22)
Mohammed Al Za'abi, Badreldin Ali, Mohammed Al Toubi
ABSTRACT
A sensitive and reproducible high-performance liquid chromatography-fluorescence method was developed and validated for quantitative determination of indoxyl sulfate in plasma using methyl paraben as an internal standard. The separation was carried out on an OSD-2 C18 Spherisorb column by isocratic elusion with sodium acetate buffer (pH 4.5) and acetonitrile (10:90, v/v) as the mobile phase. The method was validated and found to be linear in the range of 2.5 to 50 µM. The limit of quantification was 2.0 µM. The variations for intra-day and inter-day precision were less than 10.1%, and the accuracy values were between 93.4 and 102.5%. The extraction recoveries were more than 89%.
Product Description
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Methyl 4-hydroxybenzoate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)