MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Predictors and variability of urinary paraben concentrations in men and women, including before and during pregnancy.
Predictors and variability of urinary paraben concentrations in men and women, including before and during pregnancy.
Environmental health perspectives (2012-06-23)
Kristen W Smith, Joe M Braun, Paige L Williams, Shelley Ehrlich, Katharine F Correia, Antonia M Calafat, Xiaoyun Ye, Jennifer Ford, Myra Keller, John D Meeker, Russ Hauser
ABSTRACT
Parabens are suspected endocrine disruptors and ubiquitous preservatives used in personal care products, pharmaceuticals, and foods. No studies have assessed the variability of parabens in women, including during pregnancy. We evaluated predictors and variability of urinary paraben concentrations. We measured urinary concentrations of methyl (MP), propyl (PP), and butyl paraben (BP) among couples from a fertility center. Mixed-effects regression models were fit to examine demographic predictors of paraben concentrations and to calculate intraclass correlation coefficients (ICCs). Between 2005 and 2010, we collected 2,721 spot urine samples from 245 men and 408 women. The median concentrations were 112 µg/L (MP), 24.2 µg/L (PP), and 0.70 µg/L (BP). Urinary MP and PP concentrations were 4.6 and 7.8 times higher in women than men, respectively, and concentrations of both MP and PP were 3.8 times higher in African Americans than Caucasians. MP and PP concentrations were slightly more variable in women (ICC = 0.42, 0.43) than men (ICC = 0.54, 0.51), and were weakly correlated between partners (r = 0.27-0.32). Among 129 pregnant women, urinary paraben concentrations were 25-45% lower during pregnancy than before pregnancy, and MP and PP concentrations were more variable (ICCs of 0.38 and 0.36 compared with 0.46 and 0.44, respectively). Urinary paraben concentrations were more variable in women compared with men, and during pregnancy compared with before pregnancy. However, results for this study population suggest that a single urine sample may reasonably represent an individual's exposure over several months, and that a single sample collected during pregnancy may reasonably classify gestational exposure.
Product Description
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Supelco
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyl 4-hydroxybenzoate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, SAJ first grade, ≥98.0%