MilliporeSigma
  • Topical steroid and non-steroidal anti-inflammatory drugs inhibit inflammatory cytokine expression on the ocular surface in the botulinum toxin B-induced murine dry eye model.
Topical steroid and non-steroidal anti-inflammatory drugs inhibit inflammatory cytokine expression on the ocular surface in the botulinum toxin B-induced murine dry eye model.
Molecular vision (2012-07-21)
Lei Zhu, Cheng Zhang, Roy S Chuck
ABSTRACT
To evaluate the effect of the topical steroid, fluorometholone, and the non-steroidal anti-inflammatory drugs (NSAIDs), nepafenac and ketorolac, on inflammatory cytokine expression of the ocular surface in the botulium toxin B-induced murine dry eye model. Topical artificial tears (0.5% carboxymethylcellulose sodium), 0.1% fluorometholone, 0.1% nepafenac, and 0.4% ketorolac were applied 3 times per day in a dry eye mouse model 1 week after intralacrimal botulium toxin B (BTX-B) or saline (sham) injection. Tear production and corneal fluorescein staining were evaluated in all groups before injection at baseline and at 3 time points up to 4 weeks after injection. The pro-inflammatory cytokines interleukin-1β (IL-1β) and tumor necrosis factor-α (TNF-α) were evaluated by immunohistochemistry. BTX-B-injected mice showed significantly decreased aqueous tear production and increased corneal fluorescein staining at the 1 and 2 week time points compared with normal control and saline-injected mice. In the BTX-B-injected mice, immunofluorescent staining for TNF-α and IL-1β in corneal and conjunctival epithelial cells increased significantly at the 2 and 4 week time points compared to that of normal and saline-injected mice, and returned to normal levels at the 4 week time point. Topical fluorometholone significantly improved corneal surface staining in the BTX-B-injected mice after 1 week of treatment, and increased the tear production within 2 weeks, but without statistical significant difference. Topical fluorometholone significantly decreased the staining of TNF-α and IL-1β in corneal and conjunctival epithelia after 1-week treatment. Topical artificial tears, 0.1% nepafenac, and 0.4% ketorolac did not show obvious effects on tear production, corneal surface staining, and levels of IL-1β and TNF-α expression in normal, and BTX-B-injected dry eye mice. Topical fluorometholone caused suppression of inflammatory cytokine expression on the ocular surface in the Botulium toxin B-induced murine dry eye model, while topical NSAIDs demonstrated no clearly beneficial effects.
Sigma-Aldrich
Fluorometholone, ≥98%
Pricing and availability is not currently available.