MilliporeSigma
  • Aesculin inhibits matrix metalloproteinase-9 expression via p38 mitogen activated protein kinase and activator protein 1 in lipopolysachride-induced RAW264.7 cells.
Aesculin inhibits matrix metalloproteinase-9 expression via p38 mitogen activated protein kinase and activator protein 1 in lipopolysachride-induced RAW264.7 cells.
International immunopharmacology (2012-08-04)
Hee-Jung Choi, Tae-Wook Chung, Jai-Eun Kim, Han-Sol Jeong, Myungsoo Joo, Jaeho Cha, Cheorl-Ho Kim, Ki-Tae Ha
ABSTRACT
Expression of matrix metalloproteinase 9 (MMP-9) may contribute to inflammatory conditions such as arthritis, hepatitis, atherosclerosis, and pulmonary fibrosis, which involves the destruction of the extracellular matrix (ECM). Macrophages stimulated with lipopolysaccharide (LPS) express MMP-9 through the nuclear factor-kappa B (NF-κB) and activator protein 1 (AP-1) signaling pathways. Aesculin, a 6,7-dihydroxycoumarin-6-O-beta-glucopyranoside, has been highlighted for its anti-hepatotoxic, hypouricemic, antioxidative, photo-protective, and anti-apoptotic properties. In this study, we investigated the effects of aesculin on LPS-stimulated MMP-9 production and its regulatory mechanism by using murine macrophage RAW264.7 cells. Aesculin did not trigger any significant cytotoxic effect on RAW264.7 cells at concentration up to 150 μM. Secretion and expression levels of MMP-9, which were highly elevated by LPS treatment, were reduced by the addition of aesculin in a dose-dependent manner. However, gelatinolytic activity of MMP-9 was not reduced by aesculin. Luciferase activity assays and electrophoretic mobility shift assays using RAW264.7 cells showed that the inhibition of MMP-9 expression by aesculin was mediated by AP-1 rather than NF-κB. In addition, aesculin inhibited phosphorylation of p38 MAPK and subsequent activation of c-fos, a component of AP-1 transcription factor, but not JNK, ERK1/2, and c-jun. These findings suggest that aesculin is a potent drug candidate that protects against the inflammatory destruction of ECM.
Sigma-Aldrich
Esculin hydrate, ≥98.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Esculin hydrate, ≥98%
Pricing and availability is not currently available.