MilliporeSigma
  • Housefly (Musca domestica L.) control potential of Cymbopogon citratus Stapf. (Poales: Poaceae) essential oil and monoterpenes (citral and 1,8-cineole).
Housefly (Musca domestica L.) control potential of Cymbopogon citratus Stapf. (Poales: Poaceae) essential oil and monoterpenes (citral and 1,8-cineole).
Parasitology research (2012-09-08)
Peeyush Kumar, Sapna Mishra, Anushree Malik, Santosh Satya
ABSTRACT
In spite of being a major vector for several domestic, medical, and veterinary pests, the control aspect of the common housefly, Musca domestica L. (Diptera: Muscidae) is often neglected. In the present study, the essential oil of Cymbopogon citratus and its major components were evaluated for control of housefly. The chemical composition analysis of C. citratus oil by gas chromatographic mass spectrometry (GC-MS) revealed citral (47 %) and 1,8-cineole (7.5 %) as principal components. The analysis of oil vapor by solid phase microextraction (SPME/GC-MS) showed increase in citral (74.9 %) and 1,8-cineole (8.6 %) content. Assay of oil against housefly larvae and pupae through contact toxicity assay showed lethal concentration (LC)(50) value of 0.41 μl/cm(2) and of percentage inhibition rate (PIR) of 77.3 %, respectively. Fumigation assay was comparatively more effective with LC(50) of 48.6 μl/L against housefly larvae, and a PIR value of 100 % against housefly pupae. The monoterpenes, citral, and 1,8-cineole, when assessed for their insecticidal activity against housefly larvae, showed LC(50) of 0.002 and 0.01 μl/cm(2) (contact toxicity assay) and LC(50) of 3.3 and 2.4 μl/L (fumigation assay). For pupicidal assay, both citral and 1,8-cineole had a PIR value of 100 %. High efficacy of citral and 1,8-cineole against housefly, established them to be an active insecticidal agent of C. citratus oil. The study demonstrates potentiality of C. citratus oil as an excellent insecticide for housefly control, and the results open up the opportunity of oil/monoterpenes being developed into an eco-friendly, economical, and acceptable product.
Product Description
Supelco
Eucalyptol, analytical standard
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Eucalyptol, 99%
Cineole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Citral, 95%
Sigma-Aldrich
Citral, natural, ≥96%, FCC, FG
Supelco
1,8-Cineole, primary reference standard
Sigma-Aldrich
Citral, mixture of cis and trans, ≥96%, FG