MilliporeSigma
  • PEG-co-PCL nanoparticles modified with MMP-2/9 activatable low molecular weight protamine for enhanced targeted glioblastoma therapy.
PEG-co-PCL nanoparticles modified with MMP-2/9 activatable low molecular weight protamine for enhanced targeted glioblastoma therapy.
Biomaterials (2012-10-17)
Guangzhi Gu, Huimin Xia, Quanyin Hu, Zhongyang Liu, Mengyin Jiang, Ting Kang, Deyu Miao, Yifan Tu, Zhiqing Pang, Qingxiang Song, Lei Yao, Hongzhan Chen, Xiaoling Gao, Jun Chen
ABSTRACT
By taking advantage of the dramatically upregulated expression of matrix metalloproteinases MMP-2 and MMP-9 in glioblastomas and the powerful transport ability of low molecular weight protamine (LMWP), we constructed an activatable low molecular weight protamine (ALMWP) and conjugated it to PEG-PCL nanoparticles (NP) to develop a 'smart' drug delivery system for enhanced targeted glioblastoma therapy. Important parameters such as particle size distribution, zeta potential and surface content were determined, which confirmed the conjugation of ALMWP to the surface of nanoparticle. ALMWP-NP loaded with paclitaxel (PTX) exhibited a desirable pharmacokinetic and biodistribution profiles for anti-glioblastoma drug delivery. Cellular experiments showed that ALMWP-NP exhibited significantly elevated MMP-dependent cellular accumulation in C6 cells via lipid raft-mediated endocytosis and energy-dependent macropinocytosis, and improved the cytotoxicity of PTX. In vitro C6 tumor spheroid uptake confirmed the tumor penetrating ability of ALMWP-NP, in vivo imaging and glioma distribution justified its specific accumulation in the glioma. The improved glioma-targeting and tumor penetration led to an anticipated enhanced in vivo anti-glioblastoma effect: animals (nude mice bearing intracranial C6 glioma) treated with ALMWP-NP-PTX survive significantly longer than those treated with saline, Taxol(®) NP-PTX and LMWP-NP-PTX. The findings here offered strong evidence for the glioblastoma-targeting therapy of ALMWP-NP-PTX, and could also lead to a significant advancement in the application of CPPs for targeted therapy of glioma.
Sigma-Aldrich
Ethylene oxide solution, 2.5-3.3 M in THF
Pricing and availability is not currently available.
Supelco
Ethylene oxide solution, certified reference material, 50 mg/mL in methanol
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene oxide, ≥99.5%
Pricing and availability is not currently available.