MilliporeSigma
Flavin conjugates for delivery of peptide nucleic acids.
Chembiochem : a European journal of chemical biology (2012-11-07)
Fanny Marlin, Philippe Simon, Stéphanie Bonneau, Patrizia Alberti, Céline Cordier, Charlotte Boix, Loïc Perrouault, Aurélie Fossey, Tula Saison-Behmoaras, Marc Fontecave, Carine Giovannangeli
ABSTRACT
Oligonucleotides and their analogues, such as peptide nucleic acids (PNAs), can be used in chemical strategies to artificially control gene expression. Inefficient cellular uptake and inappropriate cellular localization still remain obstacles in biological applications, however, especially for PNAs. Here we demonstrate that conjugation of PNAs to flavin resulted in efficient internalization into cells through an endocytic pathway. The flavin-PNAs exhibited antisense activity in the sub-micromolar range, in the context of a treatment facilitating endosomal escape. Increased endosomal release of flavin conjugates into the cytoplasm and/or nucleus was shown by chloroquine treatment and also--when the flavin-PNA was conjugated to rhodamine, a mild photosensitizer--upon light irradiation. In conclusion, an isoalloxazine moiety can be used as a carrier and attached to a cargo biomolecule, here a PNA, for internalization and functional cytoplasmic/nuclear delivery. Our findings could be useful for further design of PNAs and other oligonucleotide analogues as potent antisense agents.
Product Description
Supelco
DNOC, PESTANAL®, analytical standard
Pricing and availability is not currently available.