MilliporeSigma
  • Ring-opening polymerization of L-lactide catalyzed by calcium complexes.
Ring-opening polymerization of L-lactide catalyzed by calcium complexes.
Dalton transactions (Cambridge, England : 2003) (2012-11-28)
Mon-Wei Hsiao, Chu-Chieh Lin
ABSTRACT
A series of calcium complexes supported by NNO-tridentate ketiminate ligands have been synthesized and characterized. X-ray structural studies indicate that these complexes bear a mononuclear feature with hexa-coordinated calcium centers bonding to two ketiminate ligands. Polymerization of L-lactide catalyzed by these complexes proceeds rapidly and well controlled in the presence of a variety of alcohols. In addition, these complexes reveal good catalytic activity even with the addition of up to 80 equiv. of benzyl alcohol as a transfer agent. Furthermore, kinetic studies show ROP of L-lactide is a first-order dependency on [LA] and a second-order dependency on [BnOH].
Sigma-Aldrich
Benzyl alcohol, SAJ first grade, ≥98.5%
Pricing and availability is not currently available.
Sigma-Aldrich
3,6-Dimethyl-1,4-dioxane-2,5-dione, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Pricing and availability is not currently available.
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Benzyl alcohol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Benzyl alcohol, SAJ special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.