MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Anti-UV activity of lignin-carbohydrate complex and related compounds.
In vivo (Athens, Greece) (2012-12-15)
Toshiyuki Nanbu, Jun Shimada, Masahiko Kobayashi, Kumi Hirano, Teho Koh, Mamoru Machino, Hirokazu Ohno, Masaji Yamamoto, Hiroshi Sakagami
ABSTRACT
We recently reported that an alkaline extract of the leaves of Sasa senanensis Rehder (SE) and Lentinus edodes mycelia extract (LEM), exhibiting lignin-carbohydrate complex (LCC)-like activity, protected cells from UV-induced injury (referred to as anti-UV activity). We investigated whether LCC is the major active components responsible for anti-UV activity. Human oral squamous cell carcinoma HSC-2 cells were exposed to short UV irradiation in phosphate-buffered saline, containing different concentrations of LCC. After culturing for 48 h in fresh culture medium, the viable cell number was determined by the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) method. From the dose-response curve, the 50% cytotoxic concentration (CC(50)) and the concentration that increased the viability of the UV-irradiated cells to 50% of the control value (EC(50)) were determined. The selectivity index (SI) was determined by the following equation: SI=CC(50)/EC(50). LCCs (Fr. VI) of pine cones and seed shell, and sulfated LCC exhibited relatively high anti-UV activity (SI=7.1-38), compared with that of SE and LEM. LCCs with lower lignin content (Fr. VII) exhibited anti-UV activity, approximately one half that of Fr. VI. However, polysaccharides (laminarin, pullulan, dextran) introduced with dimethylaminoethyl- or sulfate groups with different substitution ratios were totally inactive (SI<1). The introduction of a sulfate group to LCC did not enhance the anti-UV activity of LCC. Sodium ascorbate and vanillin were the most active (SI=65), whereas gallic acid (SI=5), epigallocatechin gallate (SI=2.6), ar-trumeron (SI<1), and turmeric extract (SI<1) were much less active. The prominent anti-UV activity of SE and LEM seems to be generated by LCCs present in the extract.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG