MilliporeSigma
  • [Role of biocompatible adhesives in the increase of rhizobia physiological activity and productivity of soybean-rhizobia symbiosis].
[Role of biocompatible adhesives in the increase of rhizobia physiological activity and productivity of soybean-rhizobia symbiosis].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2013-01-09)
L V Titova, I S Brovko, N O Leonova, S K Votselko, G A Iutinskaia, V F Patyka
ABSTRACT
On the basis of natural exopolysaccharide xanthan and exopolyacrylamide the sticky-gene composition has been developed. Addition of that composition to the culture medium provided a 26.3 times higher viability of Bradyrhizobium japonicum UCM B-6035 cells during its storage. Introduction of plant growth regulators biosil or ivin into this composition increased the survival of rhizobia. Application of gel inoculant B. japonicum favored more intensive growth of rhizosphere microorganisms, nutrient's accumulation in the soil and increased productivity of soybean-Rhizobium symbiosis.
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
Pricing and availability is not currently available.
Sigma-Aldrich
Polyacrylamide, average Mn 150,000
Pricing and availability is not currently available.