MilliporeSigma
  • Development and evaluation of a novel microemulsion formulation of elacridar to improve its bioavailability.
Development and evaluation of a novel microemulsion formulation of elacridar to improve its bioavailability.
Journal of pharmaceutical sciences (2013-01-22)
Ramola Sane, Rajendar K Mittapalli, William F Elmquist
ABSTRACT
The study objective was to develop a formulation of elacridar to overcome its dissolution-rate-limited bioavailability. Elacridar is a P-glycoprotein (P-gp) and breast cancer resistance protein (BCRP) inhibitor that has been used to improve the brain distribution of drugs that are substrates of P-gp and BCRP. The chronic use of elacridar is restricted because of the poor solubility leading to poor oral bioavailability. A microemulsion formulation using Cremophor EL, Carbitol, and Captex 355 (6:3:1) was developed. The elacridar microemulsion was effective in the inhibition of P-gp and Bcrp in Madin-Darby canine kidney II-transfected cells. Friend Leukemia Virus Strain B (FVB) mice were used to determine the bioavailability of elacridar after a 10 mg/kg dose of elacridar in the microemulsion, intraperitoneally (i.p.) and orally (p.o.); and the absolute bioavailability was determined to be 1.3 and 0.47, respectively. Coadministration of elacridar microemulsion i.p. with p.o. erlotinib in FVB mice improved the erlotinib brain penetration threefold. The current study shows that a microemulsion formulation of elacridar is effective in improving the bioavailability of elacridar and is an effective inhibitor of P-gp and Bcrp, in vitro and in vivo. It offers an alternative to the suspension and allows a decrease in the dose required to achieve a significant inhibitory effect at the blood-brain barrier.
Sigma-Aldrich
Elacridar, ≥98% (HPLC)
Pricing and availability is not currently available.