MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Pleiotropic roles of uvrY on biofilm formation, motility and virulence in uropathogenic Escherichia coli CFT073.
Pleiotropic roles of uvrY on biofilm formation, motility and virulence in uropathogenic Escherichia coli CFT073.
PloS one (2013-02-06)
Arindam Mitra, Senthilkumar Palaniyandi, Christopher D Herren, Xiaoping Zhu, Suman Mukhopadhyay
ABSTRACT
Urinary tract infections primarily caused by uropathogenic strains of Escherichia coli (E. coli) remain a significant public health problem in both developed and developing countries. An important virulence determinant in uropathogenesis is biofilm formation which requires expression of fimbriae, flagella, and other surface components such as lipopolysaccharides. In this study, we explored the regulation of uvrY and csrA genes in biofilm formation, motility and virulence determinants in uropathogenic E. coli. We found that mutation in uvrY suppressed biofilm formation on abiotic surfaces such as polyvinyl chloride, polystyrene and glass, and complementation of uvrY in the mutant restored the biofilm phenotype. We further evaluated the role of uvrY gene in expression of type 1 fimbriae, an important adhesin that facilitates adhesion to various abiotic surfaces. We found that phase variation of type 1 fimbriae between fimbriated and afimbriated mode was modulated by uvrY at its transcriptional level. Deletion mutant of uvrY lowered expression of fimbrial recombinase genes, such as fimB, fimE, and fimA, a gene encoding major fimbrial subunit. Furthermore, transcription of virulence specific genes such as papA, hlyB and galU was also reduced in the deletion mutant. Swarming motility and expression of flhD and flhC was also diminished in the mutant. Taken together, our findings unravel a possible mechanism in which uvrY facilitates biofilm formation, persistence and virulence of uropathogenic E. coli.
Sigma-Aldrich
Crystal Violet, SAJ first grade
Sigma-Aldrich
Crystal Violet, JIS special grade
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~233,000, average Mn ~99,000
Poly(vinyl chloride), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Poly(vinyl chloride), low molecular weight
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~62,000, average Mn ~35,000
Supelco
Poly(vinyl chloride), Selectophore, high molecular weight
Supelco
Poly(vinyl chloride), analytical standard, average Mw 85,000 (Typical), average Mn 40,000 (Typical)
Sigma-Aldrich
Poly(vinyl chloride), high molecular weight
Sigma-Aldrich
Crystal Violet, suitable for microscopy
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, S. No.: 785
Sigma-Aldrich
Crystal Violet Solution
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Gram-Hucker′s stain solution (I)
Sigma-Aldrich
Gentian Violet solution