MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • In chemico evaluation of prohapten skin sensitizers: behavior of 2-methoxy-4-(¹³C)methylphenol in the peroxidase peptide reactivity assay (PPRA) as an alternative to animal testing.
In chemico evaluation of prohapten skin sensitizers: behavior of 2-methoxy-4-(¹³C)methylphenol in the peroxidase peptide reactivity assay (PPRA) as an alternative to animal testing.
Toxicology letters (2013-03-05)
Fabien Merckel, Elena Giménez-Arnau, G Frank Gerberick, Jean-Pierre Lepoittevin
ABSTRACT
In chemico methods, based on the assessment of a hapten's reactivity toward peptides, have been proposed as alternative methods for the assessment of the skin sensitizing potential of chemicals. However, even with these approaches showing promise, a major drawback is the activation of prohaptens, i.e. molecules needing a metabolic activation to become reactive and therefore sensitizing. Recently, it has been proposed to couple an enzymatic activation step based on horseradish peroxidase (HRP)/hydrogen peroxide to such peptide reactivity assays. To evaluate this approach, the behavior of 2-methoxy-4-methylphenol (2M4MP), reported as a moderate sensitizer according to the Local Lymph Node Assay (LLNA), has been investigated in this assay. To follow the reaction with the peptides and characterize more easily intermediates and adducts, the molecule was first (13)C isotopically substituted at the most probable reactive position. When 2M4MP was incubated with HRP/H2O2 in a mixture PBS (pH 7.4, 0.1M)/acetonitrile 2:1, two main products were formed deriving from the formation of a quinone methide 2M4MQ subsequently trapped by either H2O2 or H2O to form a benzylic hydroperoxide or alcohol, respectively. When nucleophiles such as GSH or a peptide containing a cysteine residue (Pep-Cys) were present in the reaction medium, the quinone methide 2M4MQ was trapped by the more nucleophilic thiol function to form thio-adducts. No modifications of 2M4MP were observed when the same reactions were carried out without HRP confirming that the activation of the molecule was enzyme related. Amino nucleophiles were shown to be far less reactive towards the quinone methide 2M4MQ with only tiny formation of adducts with lysine or arginine side chains. In addition we demonstrated that the same enzymatic activation could also take place in a microemulsion based on sodium dodecyl sulfate/tert-butanol/chloroform/buffer.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
2-Methoxy-4-methylphenol, ≥98%, FG
Sigma-Aldrich
2-Methoxy-4-methylphenol, natural, 97%, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
2-Methoxy-4-methylphenol, ≥98%