MilliporeSigma
  • DNA binding and anti-cancer activity of redox-active heteroleptic piano-stool Ru(II), Rh(III), and Ir(III) complexes containing 4-(2-methoxypyridyl)phenyldipyrromethene.
DNA binding and anti-cancer activity of redox-active heteroleptic piano-stool Ru(II), Rh(III), and Ir(III) complexes containing 4-(2-methoxypyridyl)phenyldipyrromethene.
Inorganic chemistry (2013-03-13)
Rakesh Kumar Gupta, Rampal Pandey, Gunjan Sharma, Ritika Prasad, Biplob Koch, Saripella Srikrishna, Pei-Zhou Li, Qiang Xu, Daya Shankar Pandey
ABSTRACT
The synthesis of four novel heteroleptic dipyrrinato complexes [(η(6)-arene)RuCl(2-pcdpm)] (η(6)-arene = C6H6, 1; C10H14, 2) and [(η(5)-C5Me5)MCl(2-pcdpm)] (M = Rh, 3; Ir, 4) containing a new chelating ligand 4-(2-methoxypyridyl)-phenyldipyrromethene (2-pcdpm) have been described. The complexes 1-4 have been fully characterized by various physicochemical techniques, namely, elemental analyses, spectral (ESI-MS, IR, (1)H, (13)C NMR, UV/vis) and electrochemical studies (cyclic voltammetry (CV) and differential pulse voltammetry (DPV)). Structures of 3 and 4 have been determined crystallographically. In vitro antiproliferative and cytotoxic activity of these complexes has been evaluated by trypan blue exclusion assay, cell morphology, apoptosis, acridine orange/ethidium bromide (AO/EtBr) fluorescence staining, and DNA fragmentation assay in Dalton lymphoma (DL) cell lines. Interaction of 1-4 with calf thymus DNA (CT DNA) has also been supported by absorption titration and electrochemical studies. Our results suggest that in vitro antitumor activity of 1-4 lies in the order 2 > 1 > 4 > 3.
Sigma-Aldrich
Deoxyribonucleic acid from calf thymus, Genomic, unsheared
Sigma-Aldrich
Deoxyribonucleic acid from calf thymus, Type XV, Activated, lyophilized powder
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis
Sigma-Aldrich
Porphobilinogen, powder
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from calf thymus, lyophilized powder
Sigma-Aldrich
Iridium, powder, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis