MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effect of two sulfur-containing amino acids, taurine and hypotaurine in European sea bass (Dicentrarchus labrax) sperm cryopreservation.
Effect of two sulfur-containing amino acids, taurine and hypotaurine in European sea bass (Dicentrarchus labrax) sperm cryopreservation.
Cryobiology (2013-04-16)
S Martínez-Páramo, P Diogo, M T Dinis, F Soares, C Sarasquete, E Cabrita
ABSTRACT
In the present work, taurine and hypotaurine were evaluated as potential additives to improve European sea bass (Dicentrarchus labrax) sperm quality after cryopreservation. For cryopreservation, three different extenders were used: control extender (NAM), supplemented with 1mM taurine or supplemented with 1mM hypotaurine, all of them containing 10% Me₂SO as cryoprotectant. To evaluate sperm quality of fresh and thawed sperm, motility (CASA: computer assisted sperm analysis), viability (SYBR Green/propidium iodide), lipid peroxidation (malondialdehyde level), protein oxidation (carbonyl content), glutathione peroxidase, glutathione reductase and superoxide dismutase activities and DNA fragmentation (comet assay) were quantified. The result demonstrated that 1 mM hypotaurine supplemented extender increased total motility (30.1 ± 3.2%), and that 1 mM taurine extender produced higher velocity (18.1 ± 2.6 μm/s) and linearity (46.0 ± 4.8%) than the control extender (21.8 ± 3.2%, 15.5 ± 1.3 μm/s, 41.8 ± 2.4%, respectively). Cell viability, lipid peroxidation and protein oxidation were not statistically different between treatments. Similar results were obtained for glutathione peroxidase and superoxide dismutase activities. Only glutathione reductase showed differential activity before and after freezing, increasing its activity in thawed sperm. Regarding the comet assay results, taurine and hypotaurine significantly reduced DNA fragmentation (52.8 ± 0.9% and 51.8 ± 0.9%, respectively) in comparison to the control (55.7 ± 0.8%). In conclusion, for European sea bass sperm cryopreservation, extenders supplemented with 1 mM taurine and 1 mM hypotaurine improved some parameters of sperm quality after thawing, resulting in better motility and lower DNA damage than the control, two very important factors related to fertilization success.
Sigma-Aldrich
Hypotaurine, ≥98% (TLC)
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, ≥98%, FG
SAFC
Taurine
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥99%