MilliporeSigma
  • Therapy prolongation improves outcome in multisystem Langerhans cell histiocytosis.
Therapy prolongation improves outcome in multisystem Langerhans cell histiocytosis.
Blood (2013-04-17)
Helmut Gadner, Milen Minkov, Nicole Grois, Ulrike Pötschger, Elfriede Thiem, Maurizio Aricò, Itziar Astigarraga, Jorge Braier, Jean Donadieu, Jan-Inge Henter, Gritta Janka-Schaub, Kenneth L McClain, Sheila Weitzman, Kevin Windebank, Stephan Ladisch
ABSTRACT
Langerhans cell histiocytosis (LCH)-III tested risk-adjusted, intensified, longer treatment of multisystem LCH (MS-LCH), for which optimal therapy has been elusive. Stratified by risk organ involvement (high [RO+] or low [RO-] risk groups), > 400 patients were randomized. RO+ patients received 1 to 2 six-week courses of vinblastine+prednisone (Arm A) or vinblastine + prednisone + methotrexate (Arm B). Response triggered milder continuation therapy with the same combinations, plus 6-mercaptopurine, for 12 months total treatment. 6/12-week response rates (mean, 71%) and 5-year survival (84%) and reactivation rates (27%) were similar in both arms. Notably, historical comparisons revealed survival superior to that of identically stratified RO+ patients treated for 6 months in predecessor trials LCH-I (62%) or LCH-II (69%, P < .001), and lower 5-year reactivation rates than in LCH-I (55%) or LCH-II (44%, P < .001). RO- patients received vinblastine+prednisone throughout. Response by 6 weeks triggered randomization to 6 or 12 months total treatment. Significantly lower 5-year reactivation rates characterized the 12-month Arm D (37%) compared with 6-month Arm C (54%, P = .03) or to 6-month schedules in LCH-I (52%) and LCH-II (48%, P < .001). Thus, prolonging treatment decreased RO- patient reactivations in LCH-III, and although methotrexate added no benefit, RO+ patient survival and reactivation rates have substantially improved in the 3 sequential trials. (Trial No. NCT00276757 www.ClinicalTrials.gov).
Sigma-Aldrich
6-Mercaptopurine monohydrate, 98%
Pricing and availability is not currently available.