MilliporeSigma
Occupational dermatitis from dibutyl maleinate.
Contact dermatitis (1985-11-01)
J Thormann, I Hansen, J Misfeldt
ABSTRACT
10 of 20 workers in an envelope-making factory developed contact dermatitis. The cause was dibutyl maleinate, which was added to a polyvinylacetate glue. Dibutyl maleinate is a strong sensitizer and should be withdrawn from industrial use.
Sigma-Aldrich
Dibutyl maleate, 96%
Pricing and availability is not currently available.