MilliporeSigma
  • 3-Nitropyrrole and 5-nitroindole as universal bases in primers for DNA sequencing and PCR.
3-Nitropyrrole and 5-nitroindole as universal bases in primers for DNA sequencing and PCR.
Nucleic acids research (1995-07-11)
D Loakes, D M Brown, S Linde, F Hill
ABSTRACT
3-Nitropyrrole and 5-nitroindole have been assessed as universal bases in primers for dideoxy DNA sequencing and in the polymerase chain reaction (PCR). In contrast to a previous report, we have found that the introduction of more than one 3-nitropyrrole residue at dispersed positions into primers significantly reduced their efficiency in PCR and sequencing reactions. Primers containing 5-nitroindole at multiple dispersed positions were similarly affected; for both bases only a small number of substitutions were tolerated. In PCR experiments neither base, when incorporated into primers in codon third positions, was as effective as hypoxanthine, which was incorporated in six codon third positions in a 20mer oligomer. However, primers containing up to four consecutive 5-nitroindole substitutions performed well in both PCR and sequencing reactions. Consecutive 3-nitropyrrole substitutions were tolerated, but less well in comparable reactions.
Sigma-Aldrich
5-Nitroindole, 98%
Pricing and availability is not currently available.