MilliporeSigma
  • Improved direct specific determination of serum iron and total iron-binding capacity.
Improved direct specific determination of serum iron and total iron-binding capacity.
Clinical chemistry (1980-02-01)
F Ceriotti, G Ceriotti
ABSTRACT
Serum iron is released from transferrin and reduced at pH 1.7 by treating serum with a 10 g/L ascorbic acid solution in 0.1 mol/L HCl. When ferrozine is added to this reagent, it forms a complex with iron that is as intensely colored as at higher pH values, and under these conditions no turbidity is produced. The second major interference, that from copper, is eliminated by adding 1 g of thiosemicarbazide per liter, which at a low pH forms a stable, uncolored complex with copper without affecting the reaction of ferrozine with iron.
Product Description
Supelco
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, for spectrophotometric det. of Fe, 97.0%
Pricing and availability is not currently available.